Learn More
Unity Lab Services™ Biosafety Cabinet Requalification Service
Requalification Services for Biosafety Cabinet (documentation included)
Brand: Unity Lab Services™ RQBsc
This item is not returnable.
View return policy
Description
Requalification (RQ): Documented verification that equipment (as installed) is operating as intended according to manufacturers specifications.
- Verifies important equipment functions and helps ensure that the equipment operates as expected by the manufacturer and conforms to standards and requirements.
- Key tests offered based on relevant certification Standard (NSF49/en 12469/ DIN12480)
- Few examples: Filter leakage, DEHS, Air velocity, Electrical safety, Air-flow smoke pattern, KI-DISCUS.
- Please note that installation and operational qualification, ideally along with preventive maintenance and calibration, should be completed on lab equipment before requalification services can be performed.
Specifications
Biosafety Cabinet | |
Requalification Service |