Learn More
Sartorius™ Biosart™ Monitor
Microbiological and analytical filtration units offering superior performance
Brand:  Sartorius™ 16403-47-06--ACK
339.95 EUR valid until 2024-12-31
Use promo code "21897" to get your promotional price.
 View more versions of this product
Product Code. 10679792
  • 393.00 € / Pack of 48
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
The system enables the user to select the optimal hardware and consumables for their needs in quality assurance. At the heart of the system is a stainless steel manifold designed to accommodate all types of filter holder and funnels. Please note: grey filters turn black when wet. Multi-branch system for routine colony or particle counting.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
White/Black
48Pack
48/Pk.
Monitor
Sterile
48Pack
48Pack
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • X48 BIOSART100 CN G/W ,45µ IND
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose