Learn More
Thermo Scientific™ BIOShield™ 1000A Rotor Adapters
Increase rotor versatility with Thermo Scientific™ BIOShield™ 1000A Rotor Adapters, which provide support for 15/50mL conical tubes.
Brand: Thermo Scientific™ 75003642
461.58 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
Description
Increase rotor versatility with Thermo Scientific™ BIOShield™ 1000A Rotor Adapters, which provide support for 15/50mL conical tubes.
Ordering Information:A variety of other adapters (sold in sets of four unless otherwise noted) are available for use with both the BIOShield 1000A Rotor (75-003-603) and the TX-750 Swinging-Bucket Rotor with Rectangular Buckets (75-003-607/75-003-614). Contact your sales representative for more information.
Specifications
15mL Conical Tube |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.