Learn More
Thermo Scientific™ BIOShield™ 720 High Speed Swinging-Bucket Rotor
Safely spin frequently-used tubes with the Thermo Scientific™ BIOShield™ 720 High-Speed Swinging-Bucket Rotor.
Brand: Thermo Scientific™ 75003183
2638.78 EUR valid until 2024-11-30
Use promo code "21681" to get your promotional price.
This item is not returnable.
View return policy
Description
Safely spin frequently-used tubes with the Thermo Scientific™ BIOShield™ 720 High-Speed Swinging-Bucket Rotor.
Ordering Information:Tube adapters, sold in sets of four, are sold separately. Contact your sales representative for more information.
Compatible with:
Thermo Scientific™ Sorvall™ ST16, SL16, Sorvall Legend™ X1, Multifuge™ X1, Megafuge™ 16 Centrifuge Series
Specifications
BIOShield 720 Swinging Bucket Rotor w/Slide Coat anti-friction finish | |
4 x 180 mL | |
6,300 rpm | |
122 mm | |
Thermo Scientific Sorvall Legend X1, Sorvall ST 16, Multifuge X1, Megafuge 16 and SL 16 Centrifuges | |
Biocontainment certified by CAMR™, HPA, Porton Down, U.K. |
Swinging Bucket Rotor | |
7,188 x g | |
162 mm | |
50 mm | |
90° | |
Yes |