missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BIOShield™ 720 Rotor Tube Adapters

Enhance your rotor's versatility with Thermo Scientific™ BIOShield™ 720 Tube Adapters, which support a wide assortment of tube sizes.

Brand:  Thermo Scientific™ 75003678

 View more versions of this product

Product Code. 10597004

  • 554.76 € / Pack of 4

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Enhance your rotor's versatility with Thermo Scientific™ BIOShield™ 720 Tube Adapters, which support a wide assortment of tube sizes. These adapters are for use with the BIOShield 720 High Speed Swinging-Bucket Rotor.

  • Maximum tube capacity: 4 × 150mL
  • Maximum O.D. × L: 50 × 122mm
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Rotor Tube Adapter
15 mL
15 mL
Conical
15mL Conical
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.