Learn More
Agilent BioTek Accessories For MultiFlo FX Microplate Dispenser RAD peri cassette
GREENER_CHOICE
Accurate and precise peristaltic pump dispensing.
Brand:  Agilent BioTek 1260020
438.90 EUR valid until 2024-09-27
Use promo code "21716" to get your promotional price.
 View more versions of this product
Product Code. 16522102
  • 462.00 € / Each
Estimated Shipment: 18-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
For use with MultiFlo FX Microplate Dispenser RAD peri cassette
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 to 3000 μL
MultiFlo FX Microplate Dispenser
10 μL
Plastic, with large bore