missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Biotin-geldanamycin

Brand:  Novus Biologicals™ NB110-81560

Product Code. 18229910

  • 432.00 € / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Store at -20°. Avoid freeze/thaw cycles.
Synthetic Geldanamycin-Biotin 98% Pure. Purple Solid. Soluble in DMSO.
Biotin
1 mg
Geldanamycin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers