missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Biotin XX Tyramide SuperBoost™ Kit, Goat anti-mouse IgG

Biotin XX Tyramide SuperBoost™ Kit features biotin XX tyramide with poly-HRP-conjugated goat anti-mouse IgG secondary antibody.

Brand:  Invitrogen™ B40911

356.40 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 15597505

  • 540.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

SuperBoost™ tyramide signal amplification is most sensitive method for detection of low abundant targets in multiplexable fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC ), and in situ hybridization (ISH).
Features:
  • High sensitivity for detection of low-level or hard-to-detect targets by fluorescent imaging
  • Simple protocol and detection using standard filters
  • Suitable for high-resolution multiplex images, co-label with DAPI, secondary antibodies, and other SuperBoost kits
  • Requires 10-100 times less primary antibody then standard ICC/IHC/ISH experiments
Benefits:
  • Poly-HRP enhancement: Several HRP enzymes are conjugated with short polymers, enhancing signal by several fold over regular HRP systems
  • Reaction stop solution: Include an HRP stop solution to halt the HRP reaction
  • Reduction of background: Include blockers for elimination or reduction of endogenous peroxidase and fluorescent background signals
Shelf Life:
  • Stable for 6 months after receiving
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2°C to 8°C
Blocking buffer (1X): 22.5mL, Poly-HRP-conjugated goat anti-mouse secondary antibody (1X): 22.5mL, Biotin-XX tyramide reagent, Hydrogen peroxide (stabilized 3% solution): 28.5mL, Reaction buffer (20X): 6mL, Reaction stop solution: 8mg, Dimethylsulfoxide (
SuperBoost™ Kit
Biotin-XX
150 slides
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.