missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Biotrak™ Leukotriene C4/D4/E4 Assay

Enzyme immunoassay suitable for plasma and cell/tissue preparations

Brand:  Cytiva RPN224

Product Code. 10650806

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Range: 15-960pg/mL (0.75-48pg/well)
  • Sensitivity: 10pg/mL (0.5pg/well)
  • 5.5-hour protocol

TRUSTED_SUSTAINABILITY
Specifications

Specifications

15 to 960pg/mL
Each
10 pg/mL
-15°C to -30°C
Leukotriene C4/D4/E4 Biotrak assay
Microplate (12 x 8-well strip coated with goat Anti-rat IgG), Peroxidase Conjugate (lyophilized), Leukotriene C4 standard, antiserum (lyophilized), TMB Substrate, Assay Buffer concentrate, Wash Buffer concentrate
Plasma, Cell or Tissue Preparations
5.5 hr.
For Plasma and cell/tissue preparations
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

SDS
Documents

Documents

Certificates
Special Offers

Special Offers