missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)calcium(II), 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11338718
Click to view available options
Quantity:
0.5 g
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118448-18-3
Molecular Formula C22H38CaO4
Molecular Weight (g/mol) 406.62
MDL Number MFCD00064757
InChI Key DOOFPPIHJGRIGW-OLHSYEKISA-L
Synonym ca tmhd 2, calcium bis dipivaloylmethanate, bis 2,2,6,6-tetramethyl-3,5-heptanedionato calcium
PubChem CID 14598209
IUPAC Name calcium;(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate;(E)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
SMILES CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].[Ca+2]

Specifications

Melting Point 202°C to 206°C
Boiling Point 205°C (0.1 mmHg)(sublimation)
Quantity 2 g
Sensitivity Hygroscopic
Formula Weight 406.57
Percent Purity 99.99%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)calcium(II)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.