missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II), 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11384068
Click to view available options
Quantity:
0.5 g
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Precursor for CVD process

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II) is used in nano-material, thin films and ferroelectric metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13986-53-3
Molecular Formula C22H38CoO4
Molecular Weight (g/mol) 425.48
MDL Number MFCD00233616
InChI Key KLJJOSZRALJWDS-UHFFFAOYSA-N
Synonym bis 2,2,6,6-tetramethyl-3,5-heptanedionato cobalt ii, cobalt 2+ bis 2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
PubChem CID 57369563
IUPAC Name cobalt(2+);2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
SMILES [Co++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C

Specifications

Melting Point 143°C
Quantity 0.5 g
Solubility Information Soluble in hydrocarbons,alcohols and ethers. Slightly soluble in water.
Formula Weight 425.46
Percent Purity 99.9%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.