missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-benzamidophenyl) disulfide, 97%, Thermo Scientific Chemicals

Product Code. 11398615
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-57-9
Molecular Formula C26H20N2O2S2
Molecular Weight (g/mol) 456.578
MDL Number MFCD00043806
InChI Key ZHMIOPLMFZVSHY-UHFFFAOYSA-N
Synonym bis 2-benzamidophenyl disulfide, peptazin bafd, peptisant 1o, pepton 22, 2,2'-dibenzoylaminodiphenyl disulfide, bis o-benzamidophenyl disulfide, o,o'-dibenzamidodiphenyl disulfide, 2,2'-dibenzamidodiphenyl disulfide, n,n'-dithiodi-o-phenylenedibenzamide, 2',2'-dithiodibenzanilide
PubChem CID 67271
IUPAC Name N-[2-[(2-benzamidophenyl)disulfanyl]phenyl]benzamide
SMILES C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2SSC3=CC=CC=C3NC(=O)C4=CC=CC=C4

Specifications

Melting Point 140°C to 143°C
Quantity 25 g
Beilstein 721443
Formula Weight 456.59
Percent Purity 97%
Chemical Name or Material Bis(2-benzamidophenyl) disulfide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'