missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) sebacate, 97%, Thermo Scientific Chemicals

Product Code. 11925021
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-62-3
Molecular Formula C26H50O4
Molecular Weight (g/mol) 426.68
MDL Number MFCD00009497
InChI Key VJHINFRRDQUWOJ-UHFFFAOYNA-N
Synonym bis 2-ethylhexyl sebacate, bisoflex, bis 2-ethylhexyl decanedioate, plexol, bisoflex dos, monoplex dos, octoil s, reolube dos, staflex dos, di 2-ethylhexyl sebacate
PubChem CID 31218
IUPAC Name bis(2-ethylhexyl) decanedioate
SMILES CCCCC(CC)COC(=O)CCCCCCCCC(=O)OCC(CC)CCCC

Specifications

Acidity 0.15mg KOH/g max.
Melting Point -55°C
Color Yellow
Density 0.9100g/mL
Boiling Point 212°C (1.0 mmHg)
Flash Point 215°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic drum
Refractive Index 1.4500 to 1.455
Quantity 10 L
Saponification Value 260 to 265mg KOH/g
Beilstein 02,181
Residue after Evaporation 12% (10g, 6 hrs, 200°C) max.
Merck Index 15,1255
Specific Gravity 0.91
Solubility Information Solubility in water: insoluble
Viscosity ca 23 mPa.s (20°C)
Formula Weight 426.67
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Bis(2-ethylhexyl) sebacate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'