missing translation for 'onlineSavingsMsg'
Learn More
Bis(acetonitrile)(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, 97+%, Thermo Scientific Chemicals Product Code.: 15410097

Bis(acetonitrile)(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, 97+%, Thermo Scientific Chemicals

Product Code. 15410097
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15410097

Brand: Thermo Scientific Alfa Aesar H66191.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32679-02-0
Molecular Formula C12H18BF4N2Rh
Molecular Weight (g/mol) 380.00
MDL Number MFCD06798085
InChI Key KSMAFOANQHYEJG-SUESZSCISA-N
Synonym bis acetonitrile 1,5-cyclooctadiene rhodium 1 tetrafluoroborate, bis acetonitrile 1,5-cyclooctadiene rhodium i tetrafluoroborate, rh cod mecn 2 bf4, bis acetonitrile 1,5-cyclooctadiene rhodium i teShow MoreShow Less
PubChem CID 11132655
SMILES [Rh+].CC#N.CC#N.F[B-](F)(F)F.C1C\C=C/CC\C=C/1

Specifications

Color Yellow
Quantity 5 g
Sensitivity Air Sensitive
IUPAC Name acetonitrile;(1Z,5Z)-cycloocta-1,5-diene;rhodium;tetrafluoroborate
Formula Weight 379.99
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material Bis(acetonitrile)(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ19A028 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberH66191.06,H66191.03
Lot NumberX10F054 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberH66191.06,H66191.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bis(acetonitrile)(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.