missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclopentadienyl)cobalt, Thermo Scientific Chemicals

Product Code. 11363307
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Bis(cyclopentadienyl)cobalt is used to catalyze the preparation of pyridines from alkynes and nitriles. It acts as a polymerization inhibitor of olefins. It is a common one-electron reducing agent in the laboratory. It reacts with carbon monoxide to prepare cobalt(I) derivative, cyclopentadienylcobalt dicarbonyl. It also serves as an internal standard in cyclic voltametry. Further, it finds use in Diels-Alder reactions. In addition to this, it is used as a paint drier and as an oxygen stripping agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1277-43-6
Molecular Formula C10H10Co
Molecular Weight (g/mol) 189.123
MDL Number MFCD00013749
InChI Key SNBRJOIYNQIWSZ-UHFFFAOYSA-N
Synonym Cobaltocene
PubChem CID 24942376
IUPAC Name cobalt(2+);cyclopenta-1,3-diene
SMILES C1C=CC=[C-]1.C1C=CC=[C-]1.[Co+2]

Specifications

Melting Point 176°C to 180°C (decomposition)
Quantity 1 g
UN Number UN3181
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 189.12
Physical Form Powder
Chemical Name or Material Bis(cyclopentadienyl)cobalt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'type03FormHeader'

missing translation for 'productTitle'

missing translation for 'thankYou' missing translation for 'feedbackSubmitted'