missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclopentadienyl)titanium dichloride, 99+%, Thermo Scientific Chemicals

Product Code. 11363958
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This metallocene is a common reagent in organometallic and organic synthesis. Titanocene dichloride is used to prepare titanocene pentasulfide. Used as an anticancer drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1271-19-8
Molecular Formula C10H10Cl2Ti-2
Molecular Weight (g/mol) 248.957
MDL Number MFCD00003723
InChI Key MKNXBRLZBFVUPV-UHFFFAOYSA-L
Synonym Dichlorobis(cyclopentadienyl)titanium; Titanocene dichloride
PubChem CID 124040768
IUPAC Name cyclopenta-1,3-diene;titanium(2+);dichloride
SMILES [CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Cl-].[Ti+2]

Specifications

Melting Point ∼285°C (decomposition)
Color Red
Density 1.60 g/mL
Odor Odorless
Quantity 2 g
Sensitivity Air and moisture sensitive
Merck Index 14,9482
Solubility Information Reacts in moist air and in water to form HCl . Slightly hydrolysis in water.
Formula Weight 249
Percent Purity ≥99%
Physical Form Crystalline
Chemical Name or Material Bis(cyclopentadienyl)titanium dichloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'