Learn More
Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(0), Pd 16.7%, Thermo Scientific Chemicals
Description
Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(0) is used in cross-coupling including sp2-sp3 coupling, especially Suzuki, Negishi-type, Heck, Sonogashira, and Buchwald-Hartwig coupling reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | White |
Quantity | 250 mg |
Sensitivity | Air Sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 637.18 |
Concentration or Composition (by Analyte or Components) | Pd 16.7% |
Physical Form | Crystalline |
Chemical Name or Material | Bis[di-tert-butyl (4-dimethylaminophenyl)phosphine]palladium (0) |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.