missing translation for 'onlineSavingsMsg'
Learn More
Bis(dibenzylideneacetone)palladium(0), Thermo Scientific Chemicals Product Code.: 11337547

Bis(dibenzylideneacetone)palladium(0), Thermo Scientific Chemicals

Product Code. 11337547
0.25 g
Click to view available options
Quantity:
0.25 g
1 g
5 g
This item is not returnable. View return policy

Product Code. 11337547

Brand: Thermo Scientific Alfa Aesar 012764.01

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hydrogenation, isomerization, carbonylation, oxidation, C-C bond formation.Bis(dibenzylideneacetone)palladium(0) is used as a reagent for the preparation of allylic substituted cyclopentadienes. It acts as a homogeneous catalyst as well as in the alkylation of allyl acetates by various nucleophiles. Further, it is used in Suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32005-36-0
Molecular Formula C34H28O2Pd
Molecular Weight (g/mol) 575.02
MDL Number MFCD00051942
InChI Key UKSZBOKPHAQOMP-SVLSSHOZSA-N
Synonym bis dibenzylideneacetone palladium, bis dibenzylideneacetone palladium 0, pd dba 2, 1e,4e-1,5-diphenylpenta-1,4-dien-3-one; palladium, bis dibenzyldeneacetone palladium 0, tris dibenzylideneacetone dipalladium o, palladium 0 bis dibenzylideneacetone, pubchem14428
PubChem CID 6505921
IUPAC Name (1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
SMILES [Pd].O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1.O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1

Specifications

Melting Point 135°C (decomposition)
Color Red to Brown
Quantity 0.25 g
Sensitivity Air and moisture sensitive
Solubility Information Slightly soluble in CH2Cl2,CHCl3 and benzene.Soluble in dichloromethane,chloroform,1,2-dichloroethane,acetone,acetonitrile and benzene. Insoluble in water.
Formula Weight 575
Physical Form Powder
Chemical Name or Material Bis(dibenzylideneacetone) palladium(0)

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW09I012 Certificate TypeCertificate of Analysis Date16/09/2022 Catalog Number012764.06,012764.01,012764.03
Lot NumberP25I024 Certificate TypeCertificate of Analysis Date24/05/2022 Catalog Number012764.06,012764.01,012764.03
Lot NumberP29F049 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number012764.06,012764.01,012764.03
Lot NumberS16G050 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number012764.06,012764.01,012764.03
Lot NumberZ18G013 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number012764.06,012764.01,012764.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bis(dibenzylideneacetone)palladium(0), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.