missing translation for 'onlineSavingsMsg'
Learn More

Bis(dibenzylideneacetone)palladium(0), Thermo Scientific Chemicals

33.27 € - 298.49 €

Chemical Identifiers

CAS 32005-36-0
Molecular Formula C34H28O2Pd
Molecular Weight (g/mol) 575.02
MDL Number MFCD00051942
InChI Key UKSZBOKPHAQOMP-SVLSSHOZSA-N
Synonym bis dibenzylideneacetone palladium, bis dibenzylideneacetone palladium 0, pd dba 2, 1e,4e-1,5-diphenylpenta-1,4-dien-3-one; palladium, bis dibenzyldeneacetone palladium 0, tris dibenzylideneacetone dipalladium o, palladium 0 bis dibenzylideneacetone, pubchem14428
PubChem CID 6505921
IUPAC Name (1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
SMILES [Pd].O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1.O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11337547
View Documents
Thermo Scientific Alfa Aesar
012764.01
0.25 g
33.27 €
0.25g
Please to purchase this item. Need a web account? Register with us today!
11317547
View Documents
Thermo Scientific Alfa Aesar
012764.03
1 g
65.22 €
1g
Please to purchase this item. Need a web account? Register with us today!
11327547
View Documents
Thermo Scientific Alfa Aesar
012764.06
5 g
298.49 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Hydrogenation, isomerization, carbonylation, oxidation, C-C bond formation.Bis(dibenzylideneacetone)palladium(0) is used as a reagent for the preparation of allylic substituted cyclopentadienes. It acts as a homogeneous catalyst as well as in the alkylation of allyl acetates by various nucleophiles. Further, it is used in Suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

32005-36-0
575.02
UKSZBOKPHAQOMP-SVLSSHOZSA-N
6505921
[Pd].O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1.O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1
C34H28O2Pd
MFCD00051942
bis dibenzylideneacetone palladium, bis dibenzylideneacetone palladium 0, pd dba 2, 1e,4e-1,5-diphenylpenta-1,4-dien-3-one; palladium, bis dibenzyldeneacetone palladium 0, tris dibenzylideneacetone dipalladium o, palladium 0 bis dibenzylideneacetone, pubchem14428
(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium

Specifications

32005-36-0
Red to Brown
MFCD00051942
bis dibenzylideneacetone palladium, bis dibenzylideneacetone palladium 0, pd dba 2, 1e,4e-1,5-diphenylpenta-1,4-dien-3-one; palladium, bis dibenzyldeneacetone palladium 0, tris dibenzylideneacetone dipalladium o, palladium 0 bis dibenzylideneacetone, pubchem14428
UKSZBOKPHAQOMP-SVLSSHOZSA-N
(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
6505921
Powder
135°C (decomposition)
C34H28O2Pd
Air and moisture sensitive
Slightly soluble in CH2Cl2,CHCl3 and benzene.Soluble in dichloromethane,chloroform,1,2-dichloroethane,acetone,acetonitrile and benzene. Insoluble in water.
[Pd].O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1.O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1
575.02
575
Bis(dibenzylideneacetone) palladium(0)
Videos
Safety and Handling

Safety and Handling

P261-P272-P280g-P302+P352-P333+P313-P363-P501c

H317

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.