missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(ethylenedithio)tetrathiafulvalene
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
missing translation for 'unitSize'
1g
250mg
Beschreibung
It is used as a component for organic superconductors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Spezifikation
Spezifikation
| Melting Point | 238°C (decomposition) |
| Quantity | 250 mg |
| Solubility Information | Soluble in organic solvents. |
| Formula Weight | 384.68 |
| Physical Form | Powder |
| Chemical Name or Material | Bis(ethylenedithiolo)tetrathiafulvalene |
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur