missing translation for 'onlineSavingsMsg'
Learn More

Bis(heptafluoroisopropyl)ketone, 97%, Thermo Scientific Chemicals

Product Code. 11339345
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 813-44-5
Molecular Formula C7F14O
Molecular Weight (g/mol) 366.054
MDL Number MFCD00042087
InChI Key GRVMOMUDALILLH-UHFFFAOYSA-N
Synonym bis perfluoroisopropyl ketone, 1,1,1,2,4,5,5,5-octafluoro-2,4-bis trifluoromethyl pentan-3-one, bis perfluorisopropyl ketone, bis heptafluoroisopropyl ketone, 3-pentanone, 1,1,1,2,4,5,5,5-octafluoro-2,4-bis trifluoromethyl, perfluoroisopropyl ketone, bis heptafluoroisopropyl methanone, cf3 2cfc o cf cf3 2, 3-hexanone, 1,1,1,2,4,4,5,5,6,6,6-undecafluoro-2-trifluoromethyl, tert-butyl 2-oxo-1-pyrrolidinecarboxylat
PubChem CID 69941
IUPAC Name 1,1,1,2,4,5,5,5-octafluoro-2,4-bis(trifluoromethyl)pentan-3-one
SMILES C(=O)(C(C(F)(F)F)(C(F)(F)F)F)C(C(F)(F)F)(C(F)(F)F)F

Specifications

Density 1.661
Boiling Point 72°C to 73°C
Quantity 1 g
UN Number UN2810
Beilstein 1916532
Formula Weight 366.06
Percent Purity 97%
Chemical Name or Material Bis(heptafluoroisopropyl)ketone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'