missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bis(methylthio)gliotoxin, 99%

Product Code. 15468749
Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Description Colorless
Content And Storage -30°C to -10°C
CAS 74149-38-5
Molecular Formula C15H20N2O4S2
Formula Weight 356.47
Synonym FR-49175;
Product Type Bis(methylthio)gliotoxin, 99%
Form Oil
InChI Key OVBAGMZLGLXSBN-UOVKNHIHSA-N
SMILES CS[C@@]12CC3=CC=C[C@H](O)[C@H]3N1C(=O)[C@@](CO)(SC)N(C)C2=O
IUPAC Name (3R,5aS,6S,10aR)-6-hydroxy-3-(hydroxymethyl)-2-methyl-3,10a-bis(methylsulfanyl)-1H,2H,3H,4H,5aH,6H,10H,10aH-pyrazino[1,2-a]indole-1,4-dione
Molecular Weight (g/mol) 356.46
Assay 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.