missing translation for 'onlineSavingsMsg'
Learn More

Bis(phenylsulfonyl)sulfide, 96%, Thermo Scientific Chemicals

Product Code. 11945741
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4388-22-1
Molecular Formula C12H10O4S3
Molecular Weight (g/mol) 314.39
MDL Number MFCD00094733
InChI Key YQUSJUJNDKUWAM-UHFFFAOYSA-N
Synonym bis phenylsulfonyl sulfide, benzenesulfonyl sulfide, benzenesulfonic thioanhydride, benzenesulfonothioic acid, anhydrosulfide, benzenesulfonic acid, thio-, anhydrosulfide, bis phenylsulfonyl thio, benzenesulfonyl sulfanylsulfonyl benzene, phso2 2s, benzenesulfonylthio sulfonylbenzene
PubChem CID 257516
IUPAC Name benzenesulfonylsulfanylsulfonylbenzene
SMILES O=S(=O)(SS(=O)(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 129.0°C to 134.0°C
Color Pink to White
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 11,III,163
Formula Weight 314.41
Percent Purity 96%
Physical Form Crystals or Powder
Chemical Name or Material Bis(phenylsulfonyl)sulfide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'