missing translation for 'onlineSavingsMsg'
Learn More
Bis(pinacolato)diboron, 98%, Thermo Scientific Chemicals Product Code.: 10544634

Bis(pinacolato)diboron, 98%, Thermo Scientific Chemicals

Product Code. 10544634
25 g, Glass bottle
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10544634

Brand: Thermo Scientific Acros 330570250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73183-34-3
Molecular Formula C12H24B2O4
Molecular Weight (g/mol) 253.94
MDL Number MFCD00799570
InChI Key IPWKHHSGDUIRAH-UHFFFAOYSA-N
Synonym bis pinacolato diboron, bis pinacolato diborane, 4,4,4',4',5,5,5',5'-octamethyl-2,2'-bi 1,3,2-dioxaborolane, pinacol diborane, diboron pinacol ester, b2pin2, 4,4,4',4',5,5,5',5'-octamethyl-2,2'-bi-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-tetramethyl-1,3,2-dioxaborolan-2-yl-1,3,2-dioxaborolane, bispinacolate diboron, unii-i906w26p4u
PubChem CID 2733548
IUPAC Name 4,4,5,5-tetramethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 135.0°C to 140.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Merck Index 15,1301
Formula Weight 253.9
Percent Purity 98%
Physical Form Powder or Platelets
Chemical Name or Material Bis(pinacolato)diboron
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0459346 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number330570010,330570250,330571000,330570050
Lot NumberA0467125 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number330570010,330570250,330571000,330570050
Lot NumberA0429379 Certificate TypeCertificate of Analysis Date26/04/2024 Catalog Number330570010,330570250,330571000,330570050
Lot NumberA0450418 Certificate TypeCertificate of Analysis Date21/02/2024 Catalog Number330570010,330570250,330571000,330570050
Lot NumberA0459346 Certificate TypeCertificate of Analysis Date19/01/2024 Catalog Number330570010,330570250,330571000,330570050,330572500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bis(pinacolato)diboron, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.