missing translation for 'onlineSavingsMsg'
Learn More
Bis(tri-n-butyltin) oxide, 97%, Thermo Scientific Chemicals Product Code.: 11490773

Bis(tri-n-butyltin) oxide, 97%, Thermo Scientific Chemicals

Product Code. 11490773
500 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11490773

Brand: Thermo Scientific Alfa Aesar A13242.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bis(tri-n-butyltin) oxide is employed in the synthesis of α,β-unsaturated methyl ketones, isoxazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(tri-n-butyltin) oxide is employed in the synthesis of α,β-unsaturated methyl ketones, isoxazoles.

Solubility
Soluble in water (0.071 g/L)

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-35-9
Molecular Formula C24H58OSn2
Molecular Weight (g/mol) 600.15
MDL Number MFCD00009418
InChI Key NZQFYJHTWVEUPP-UHFFFAOYSA-N
Synonym tributyltin oxide, tbto, bis tributyltin oxide, hexabutyldistannoxane, distannoxane, hexabutyl, lastanox q, bis tri-n-butyltin oxide, butinox, biomet
PubChem CID 16682746
ChEBI CHEBI:81543
IUPAC Name tributyl(tributylstannyloxy)stannane
SMILES O.CCCC[SnH](CCCC)CCCC.CCCC[SnH](CCCC)CCCC

Specifications

Melting Point -45°C
Density 1.172
Boiling Point 179°C to 180°C (2 mmHg)
Flash Point 168°C (334°F)
Odor Irritating
Linear Formula [(CH3CH2CH2CH2)3Sn]2O
Refractive Index 1.4865
Quantity 500 g
UN Number UN2788
Beilstein 745057
Solubility Information Soluble in water (0.071g/L)
Formula Weight 596.07
Percent Purity 97%
Chemical Name or Material Bis(tri-n-butyltin) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ26I066 Certificate TypeCertificate of Analysis Date02/05/2022 Catalog NumberA13242.0E,A13242.36,A13242.22
Lot NumberQ06I003 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA13242.0E,A13242.36,A13242.22
Lot NumberZ06H008 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA13242.0E,A13242.36,A13242.22
Lot NumberX20H035 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA13242.0E,A13242.36,A13242.22
Lot NumberD5029C Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13242.0E,A13242.36,A13242.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bis(tri-n-butyltin) oxide, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.