missing translation for 'onlineSavingsMsg'
Learn More
Bis(tri-tert-butylphosphine)palladium(0), 98%, Thermo Scientific Chemicals Product Code.: 10628922

Bis(tri-tert-butylphosphine)palladium(0), 98%, Thermo Scientific Chemicals

Product Code. 10628922
100 mg, Glass bottle
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10628922

Brand: Thermo Scientific Acros 363501000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53199-31-8
Molecular Formula C24H54P2Pd
Molecular Weight (g/mol) 511.06
MDL Number MFCD03094580
InChI Key MXQOYLRVSVOCQT-UHFFFAOYSA-N
Synonym bis tri-tert-butylphosphine palladium 0, bis tri-t-butylphosphine palladium 0, bis tri-tert-butylphosphine palladium, bis tri-tert-butylphosphane palladium, palladium; tritert-butylphosphane, palladium, bis tris 1,1-dimethylethyl phosphine, pd p tbu 3 2, pd t-bu3p 2, bis tri-t-butylphosphine palladium, di tri-tert-butylphosphine palladium 0
PubChem CID 2734558
SMILES [Pd].CC(C)(C)P(C(C)(C)C)C(C)(C)C.CC(C)(C)P(C(C)(C)C)C(C)(C)C

Specifications

Melting Point 258.0°C to 272.0°C
Color Brown-Orange to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex Pd)
Packaging Glass bottle
Quantity 100 mg
Solubility Information Solubility in water: insoluble
IUPAC Name bis(tri-tert-butylphosphane) palladium
Formula Weight 511.05
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material Bis(tri-tert-butylphosphine)palladium(0)
Show More Show Less
compliance-icons
Product Identifier
  • Bis(tri-tert-butylphosphine)palladium(0)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bis(tri-tert-butylphosphine)palladium(0), 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.