missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(tri-tert-butylphosphine)palladium(0), Thermo Scientific Chemicals
Click to view available options
Quantity:
0.25 g
1 g
Description
Bis(tri-tert-butylphosphine)palladium(0) is used in Coupling reactions, Heck couplings. It acts as a palladium catalyst for reductive carbonylation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under inert atmosphere
Specifications
Specifications
Melting Point | 258°C to 272°C |
Color | White |
Quantity | 1 g |
Sensitivity | Air and light sensitive |
Solubility Information | Soluble in toluene benzene,slight soluble in chloroform and acetone. Decomposes in water. |
Formula Weight | 511.06 |
Concentration or Composition (by Analyte or Components) | Pd 20.9% |
Physical Form | Powder |
Chemical Name or Material | Bis (tri-tert-butylphosphine)palladium (0) |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'