missing translation for 'onlineSavingsMsg'
Learn More

Bis(tricyclohexylphosphine)palladium(0), 98%

Product Code. 10531482 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 10531482

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 33309-88-5 | C36H66P2Pd | 667.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 33309-88-5
Color Green to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C36H66P2Pd
MDL Number MFCD01073796
Quantity 100 mg
Synonym bis tricyclohexylphosphine palladium 0, bis tricyclohexylphosphine palladium o, bis tricyclohexylphosphine palladium, palladium, bis tricyclohexylphosphine, pd pcy3 2, palladium; tricyclohexylphosphane, palladium-tricyclohexylphosphine 1:2, bis tricyclohexylphosphine-palladium o
Solubility Information Solubility in water: insoluble in water
InChI Key JGBZTJWQMWZVNX-UHFFFAOYSA-N
SMILES [Pd].C1CCC(CC1)P(C1CCCCC1)C1CCCCC1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1
IUPAC Name palladium;tricyclohexylphosphane
Molecular Weight (g/mol) 667.29
PubChem CID 2734559
Formula Weight 667.28
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Bis(tricyclohexylphosphine)palladium(0)
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.