missing translation for 'onlineSavingsMsg'
Learn More

Bis(triethylphosphine)palladium(II) chloride, Thermo Scientific Chemicals

Product Code. 10722104 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10722104

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28425-04-9
Molecular Formula C12H30Cl2P2Pd
Molecular Weight (g/mol) 413.64
MDL Number MFCD00191831
InChI Key ULYNIEUXPCUIEL-UHFFFAOYSA-L
Synonym dichlorobis triethylphosphine palladium ii, bis triethylphosphine palladium dichloride, trans-dichlorobis triethylphosphine palladium ii, bis triethylphosphine palladium ii chloride, pdcl2 pet3 2, palladium,dichlorobis triethylphosphine, dichloropalladium-triethylphosphane 1/2, bis triethylphosphine dichloridopalladium ii
PubChem CID 11144124
IUPAC Name dichloropalladium;triethylphosphane
SMILES Cl[Pd]Cl.CCP(CC)CC.CCP(CC)CC

Specifications

Melting Point 139°C to 142°C
Color Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 250 mg
Solubility Information Solubility in water: insoluble
Formula Weight 413.64
Physical Form Powder
Chemical Name or Material Bis(triethylphosphine)palladium(II) chloride
compliance-icons
Product Identifier
  • Bis(triethylphosphine)palladium(II) chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.