missing translation for 'onlineSavingsMsg'
Learn More

Bis(trifluoromethanesulfonyl)imide, 99%, Thermo Scientific Chemicals

Product Code. 10041593
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82113-65-3
Molecular Formula C2HF6NO4S2
Molecular Weight (g/mol) 281.16
MDL Number MFCD00214154
InChI Key ZXMGHDIOOHOAAE-UHFFFAOYSA-N
Synonym 1,1,1-trifluoro-n-trifluoromethyl sulfonyl methanesulfonamide, trifluoromethanesulfonimide, unii-c7r849vm9l, bis trifluoromethanesulfonyl imide, bistrifluoromethanesulfonimide, 1,1,1-trifluoro-n-trifluoromethanesulfonylmethanesulfonamide, 1,1,1-trifluoro-n-trifluoromethylsulfonyl methanesulfonamide, bis trifluoromethane sulfonimide, n,n-bis trifluoromethanesulfonyl imide, n,n-bis trifluoromethylsulphonyl amine
PubChem CID 157857
IUPAC Name 1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide
SMILES C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F

Specifications

Melting Point 52°C to 56°C
Color Brown to Colorless
Boiling Point 90°C to 91°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: reacts
Formula Weight 281.16
Percent Purity 99%
Physical Form Crystals
Chemical Name or Material Bis(trifluoromethanesulfonyl)imide
Show More Show Less
compliance-icons
Product Identifier
  • Bis(trifluoromethanesulfonyl)imide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.