missing translation for 'onlineSavingsMsg'
Learn More

Bis(trimethylsilyl)acetylene, 99%

Product Code. 10206050 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 10206050

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14630-40-1 | C8H18Si2 | 170.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14630-40-1
Molecular Formula C8H18Si2
Molecular Weight (g/mol) 170.41
MDL Number MFCD00008276
InChI Key ZDWYFWIBTZJGOR-UHFFFAOYSA-N
Synonym bis trimethylsilyl acetylene, silane, 1,2-ethynediylbis trimethyl, 1,2-bis trimethylsilyl ethyne, btmsa, 1,2-bis-trimethylsilanyl-ethyne, ethyne-1,2-diylbis trimethylsilane, 2,5-disilahex-3-yne, 2,2,5,5-tetramethyl, trimethyl 2-trimethylsilyl ethynyl silane, silane, 1,1'-1,2-ethynediyl bis 1,1,1-trimethyl
PubChem CID 84564
IUPAC Name trimethyl(2-trimethylsilylethynyl)silane
SMILES C[Si](C)(C)C#C[Si](C)(C)C

Specifications

Melting Point 21.0°C to 24.0°C
Color Colorless
Density 0.7500g/mL
Boiling Point 136.0°C to 137.0°C
Flash Point 2°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3SiC=CSi(CH3)3
Refractive Index 1.4260 to 1.4280
Quantity 10 g
Fieser 05,44; 13,97
Specific Gravity 0.75
Solubility Information Solubility in water: may decompose
Formula Weight 170.41
Percent Purity 99%
Physical Form Liquid After Melting
Chemical Name or Material Bis(trimethylsilyl)acetylene, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Bis(trimethylsilyl)acetylene
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.