Learn More
Bis(trimethylsilyl)methane, 98%, Thermo Scientific Chemicals
31.20 € - 435.17 €
Chemical Identifiers
CAS | 2117-28-4 |
---|---|
Molecular Formula | C7H20Si2 |
Molecular Weight (g/mol) | 160.41 |
MDL Number | MFCD00008275 |
InChI Key | GYIODRUWWNNGPI-UHFFFAOYSA-N |
Synonym | bis trimethylsilyl methane, methylenebis trimethylsilane, silane, methylenebis trimethyl, trimethyl trimethylsilylmethyl silane, trimethyl trimethylsilyl methyl silane, 2,4-disilapentane, 2,2,4,4-tetramethyl, silane, 1,1'-methylenebis 1,1,1-trimethyl |
PubChem CID | 75029 |
IUPAC Name | trimethyl(trimethylsilylmethyl)silane |
SMILES | C[Si](C)(C)C[Si](C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11427587
|
Thermo Scientific Alfa Aesar
A15662.03 |
1 g | N/A | ||||||
11437587
|
Thermo Scientific Alfa Aesar
A15662.06 |
5 g |
110.55 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11447587
|
Thermo Scientific Alfa Aesar
A15662.14 |
25 g |
435.17 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2117-28-4 | |
160.41 | |
GYIODRUWWNNGPI-UHFFFAOYSA-N | |
75029 | |
C[Si](C)(C)C[Si](C)(C)C |
C7H20Si2 | |
MFCD00008275 | |
bis trimethylsilyl methane, methylenebis trimethylsilane, silane, methylenebis trimethyl, trimethyl trimethylsilylmethyl silane, trimethyl trimethylsilyl methyl silane, 2,4-disilapentane, 2,2,4,4-tetramethyl, silane, 1,1'-methylenebis 1,1,1-trimethyl | |
trimethyl(trimethylsilylmethyl)silane |
Specifications
132°C to 133°C | |
2117-28-4 | |
MFCD00008275 | |
1734530 | |
bis trimethylsilyl methane, methylenebis trimethylsilane, silane, methylenebis trimethyl, trimethyl trimethylsilylmethyl silane, trimethyl trimethylsilyl methyl silane, 2,4-disilapentane, 2,2,4,4-tetramethyl, silane, 1,1'-methylenebis 1,1,1-trimethyl | |
C[Si](C)(C)C[Si](C)(C)C | |
160.41 | |
160.41 | |
1.417 | |
Bis(trimethylsilyl)methane |
-71°C | |
C7H20Si2 | |
UN1993 | |
12°C (53°F) | |
GYIODRUWWNNGPI-UHFFFAOYSA-N | |
trimethyl(trimethylsilylmethyl)silane | |
75029 | |
98% | |
0.75 |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 218-322-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.