missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(triphenylphosphine)palladium(II) diacetate, 99%, Thermo Scientific Chemicals
65.22 € - 263.31 €
Chemical Identifiers
CAS | 14588-08-0 |
---|---|
Molecular Formula | C40H36O4P2Pd |
Molecular Weight (g/mol) | 749.09 |
MDL Number | MFCD00010013 |
InChI Key | YOUIUHDWQIUKAO-UHFFFAOYSA-L |
Synonym | bis triphenylphosphinepalladium acetate |
PubChem CID | 73357379 |
IUPAC Name | palladium(2+);triphenylphosphanium;diacetate |
SMILES | CC(=O)O[Pd++]OC(C)=O.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10092790
|
Thermo Scientific Acros
209270010 |
1 g | Glass bottle |
65.22 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10511081
|
Thermo Scientific Acros
209270050 |
5 g | Glass bottle |
263.31 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14588-08-0 | |
749.09 | |
YOUIUHDWQIUKAO-UHFFFAOYSA-L | |
73357379 | |
CC(=O)O[Pd++]OC(C)=O.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
C40H36O4P2Pd | |
MFCD00010013 | |
bis triphenylphosphinepalladium acetate | |
palladium(2+);triphenylphosphanium;diacetate |
Specifications
14588-08-0 | |
Yellow | |
14% min. (Pd) | |
[(C6H5)3P]2Pd(O2CCH3)2 | |
bis triphenylphosphinepalladium acetate | |
CC(=O)O[Pd++]OC(C)=O.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
749.09 | |
749.08 | |
Powder |
136.0°C | |
Authentic | |
C40H36O4P2Pd | |
MFCD00010013 | |
YOUIUHDWQIUKAO-UHFFFAOYSA-L | |
palladium(2+);triphenylphosphanium;diacetate | |
73357379 | |
99% | |
Bis(triphenylphosphine)palladium(II) diacetate, 99% |
Safety and Handling
missing translation for 'einecsNumber' : 238-628-4
Product Identifier
- Bis(triphenylphosphine)palladium(II) diacetate
RUO – Research Use Only