missing translation for 'onlineSavingsMsg'
Learn More

Bismuth 2-ethylhexanoate, Thermo Scientific Chemicals

Product Code. 11374237
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Bismuth 2-ethylhexanoate is used as an industrial lubricant and dessicant. It is also used for synthesizing of urethane foam, urethane coating and other urethane products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67874-71-9
Molecular Formula C24H48BiO6
Molecular Weight (g/mol) 641.622
MDL Number MFCD00043803
InChI Key MWXWNYHYPMRCHN-UHFFFAOYSA-N
Synonym 2-Ethylhexanoic acid bismuth salt
PubChem CID 131857451
IUPAC Name bismuth;2-ethylhexanoic acid
SMILES CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.[Bi]

Specifications

Quantity 25 g
Solubility Information Not miscible with water.
Formula Weight 638.61
Physical Form Viscous Liquid
Chemical Name or Material Bismuth 2-ethylhexanoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'