missing translation for 'onlineSavingsMsg'
Learn More

Bismuth, 99%, powder, -100 mesh, Thermo Scientific Chemicals

Product Code. 10721365 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10721365

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-69-9
Molecular Formula Bi
Molecular Weight (g/mol) 208.98
MDL Number MFCD00134033
InChI Key JCXGWMGPZLAOME-UHFFFAOYSA-N
Synonym pieces, bismuth, elemental, rod, 12.7mm 0.5in dia, atom, rod, powder, unii-u015tt5i8h, iii ion, shot, elongated, needles
PubChem CID 5359367
ChEBI CHEBI:33301
IUPAC Name bismuth
SMILES [Bi]

Specifications

Color Black
Melting Point 271.0°C
Boiling Point 1500.0°C
Density 9.8000 g/mL
Quantity 500 g
Packaging Plastic bottle
Assay Percent Range 99.5%
Form Powder
Linear Formula BI
Fieser 13,39
Merck Index 15, 1261
Formula Weight 208.98
Percent Purity 99.5%
Chemical Name or Material Bismuth, 99.5%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.