missing translation for 'onlineSavingsMsg'
Learn More

Bismuth aluminum oxide hydrate, 95.5%, Thermo Scientific™

Artikelnummer. 11323978
missing translation for 'orderingAttributeHoverText'
Quantity:
50 g
250 g
1 kg
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

MDL Number MFCD00798526
Synonym Aluminum bismuth oxide hydrate; Bismuth aluminate hydrate

Spezifikation

Physical Form Powder
Quantity 250 g
Solubility Information Insoluble in water; Soluble in nitric acid; Typical tap density 0;69g/cm3
Formula Weight 771.84 (Anhydrous)
Percent Purity 95.5%
Chemical Name or Material Bismuth aluminum oxide hydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'