missing translation for 'onlineSavingsMsg'
Learn More

Bismuth citrate, 94%, Thermo Scientific Chemicals

102.51 € - 365.82 €

Chemical Identifiers

CAS 813-93-4
Molecular Formula C6H5BiO7
Molecular Weight (g/mol) 398.079
MDL Number MFCD00050817
InChI Key ANERHPOLUMFRDC-UHFFFAOYSA-K
Synonym bismuth citrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, bismuth 3+ salt 1:1, bismuth citrate usp:usp, bismuth iii citrate, bismuth citrate usp, bismuth 3+ ion citrate, bismuth, citrato 3-, bismuth iii citrate 250g, bismuth 2-hydroxypropane-1,2,3-tricarboxylate
PubChem CID 13135
IUPAC Name bismuth;2-hydroxypropane-1,2,3-tricarboxylate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.[Bi+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11363988
SDS View Documents
Thermo Scientific Alfa Aesar
040398.30
250 g
102.51 €
250g
Please to purchase this item. Need a web account? Register with us today!
11353988
SDS View Documents
Thermo Scientific Alfa Aesar
040398.A1
1 kg
365.82 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Bismuth citrate is used as a progressive hair dye in cosmetics. It is used as antimicrobial agents for syphills, colitis, wound infection, quartan malaria, dyspepsia and peptic ulcers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 212-390-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bismuth citrate, 94%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.