missing translation for 'onlineSavingsMsg'
Learn More

Bismuth hydroxide, Thermo Scientific Chemicals

Product Code. 10590574
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10361-43-0
Molecular Formula BiH3O3
Molecular Weight (g/mol) 260.00
MDL Number MFCD00046188
InChI Key TZSXPYWRDWEXHG-UHFFFAOYSA-K
IUPAC Name bismuth(3+) trihydroxide
SMILES [OH-].[OH-].[OH-].[Bi+3]

Specifications

Melting Point 415.0°C
Color White to Cream
Physical Form Powder
Quantity 50 g
Linear Formula Bi(OH)3
Merck Index 15, 1267
Solubility Information Solubility in water: negligible. Other solubilities: soluble in acids
Formula Weight 260
Identification Positive
Loss on Drying 1.0% max.
Packaging Glass bottle
Chemical Name or Material Bismuth hydroxide
Show More Show Less
Product Identifier
  • Bismuth hydroxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.