missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) bromide, 97%, Thermo Scientific Chemicals

72.76 € - 207.03 €

Chemical Identifiers

CAS 7787-58-8
Molecular Formula BiBr3
Molecular Weight (g/mol) 448.69
InChI Key TXKAQZRUJUNDHI-UHFFFAOYSA-K
Synonym bismuth tribromide, bismuth iii bromide, bismuth bromide, bismuthine, tribromo, tribromobismuthine, unii-dt59198t2x, bismuth ii bromide, tribromobismuth, bismuthine,tribromo
PubChem CID 82232
IUPAC Name tribromobismuthane
SMILES Br[Bi](Br)Br
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
13408688
SDS View Documents
Thermo Scientific Acros
447770250
25 g Glass bottle
72.76 €
25g
Please to purchase this item. Need a web account? Register with us today!
13418688
SDS View Documents
Thermo Scientific Acros
447771000
100 g Glass Bottle
207.03 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7787-58-8
448.69
bismuth tribromide, bismuth iii bromide, bismuth bromide, bismuthine, tribromo, tribromobismuthine, unii-dt59198t2x, bismuth ii bromide, tribromobismuth, bismuthine,tribromo
tribromobismuthane
BiBr3
TXKAQZRUJUNDHI-UHFFFAOYSA-K
82232
Br[Bi](Br)Br

Specifications

453°C
7787-58-8
BiBr3
15,1262
Solubility in water: reacts. Other solubilities: practically insoluble in alcohol,soluble in solutions of ki,kbr,kcl,dil hcl
Br[Bi](Br)Br
448.69
448.69
Bismuth(III) bromide
218°C
96 to 103% (Bi)
BiBr3
bismuth tribromide, bismuth iii bromide, bismuth bromide, bismuthine, tribromo, tribromobismuthine, unii-dt59198t2x, bismuth ii bromide, tribromobismuth, bismuthine,tribromo
TXKAQZRUJUNDHI-UHFFFAOYSA-K
tribromobismuthane
82232
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 232-121-1

Product Identifier
  • Bismuth(III) bromide

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.