missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) bromide, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11383317
Click to view available options
Quantity:
25 g
100 g

It catalyzes the formation of cyclic carbonates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-58-8
Molecular Formula BiBr3
Molecular Weight (g/mol) 448.692
MDL Number MFCD00014195
InChI Key TXKAQZRUJUNDHI-UHFFFAOYSA-K
Synonym bismuth tribromide, bismuth iii bromide, bismuth bromide, bismuthine, tribromo, tribromobismuthine, unii-dt59198t2x, bismuth ii bromide, tribromobismuth, bismuthine,tribromo
PubChem CID 82232
IUPAC Name tribromobismuthane
SMILES Br[Bi](Br)Br

Specifications

Boiling Point 453°C
Melting Point 218°C
Physical Form Powder
Quantity 25 g
Assay Percent Range 99% (metals basis)
Solubility Information Decomposes in water.
Formula Weight 448.71
Sensitivity Hygroscopic
Density 5.72
Chemical Name or Material Bismuth(III) bromide
compliance-icons
Product Identifier
  • Bismuth(III) bromide
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.