missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) chloride oxide, 99.95% (metals basis), Thermo Scientific Chemicals

101.51 € - 255.27 €

Chemical Identifiers

CAS 7787-59-9
Molecular Formula BiClHO
Molecular Weight (g/mol) 261.437
MDL Number MFCD00010895
InChI Key BWOROQSFKKODDR-UHFFFAOYSA-N
Synonym bismuth oxychloride, mearlite, pearl white, bismuthine, chlorooxo, wismutchloridoxid, bismut-chlorid-oxid, biju, bismuth chloride oxide, basic bismuth chloride, wismut iii-chlorid-oxid
PubChem CID 6328152
IUPAC Name oxobismuth;hydrochloride
SMILES O=[Bi].Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11323327
View Documents
Thermo Scientific Alfa Aesar
017131.14
25 g
101.51 €
25g
Please to purchase this item. Need a web account? Register with us today!
11313327
View Documents
Thermo Scientific Alfa Aesar
017131.22
100 g
255.27 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used as skin protective's, thickeners, and absorbent agents. It act as a coloring agents in cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7787-59-9
261.437
BWOROQSFKKODDR-UHFFFAOYSA-N
6328152
O=[Bi].Cl
BiClHO
MFCD00010895
bismuth oxychloride, mearlite, pearl white, bismuthine, chlorooxo, wismutchloridoxid, bismut-chlorid-oxid, biju, bismuth chloride oxide, basic bismuth chloride, wismut iii-chlorid-oxid
oxobismuth;hydrochloride

Specifications

(decomposition)
Brown
BiClHO
MFCD00010895
bismuth oxychloride, mearlite, pearl white, bismuthine, chlorooxo, wismutchloridoxid, bismut-chlorid-oxid, biju, bismuth chloride oxide, basic bismuth chloride, wismut iii-chlorid-oxid
BWOROQSFKKODDR-UHFFFAOYSA-N
oxobismuth;hydrochloride
6328152
99.95%
2.15
(metals basis)
7787-59-9
Powder
BiOCl
14,1263
Soluble in HCl,HNO3. Insoluble in water,alcohol.
O=[Bi].Cl
261.437
260.43
Odorless
7.72 g/mL
Bismuth(III) chloride oxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 232-122-7

missing translation for 'rtecsNumber' : EB2700000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Bismuth(III) chloride oxide

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.