missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) chloride oxide, 99.999% (metals basis)

Product Code. p-7048882
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11361908

Brand: Thermo Scientific Alfa Aesar 011843.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7787-59-9 | BiClHO | 261.437 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-59-9
Molecular Formula BiClHO
Molecular Weight (g/mol) 261.437
MDL Number MFCD00010895
InChI Key BWOROQSFKKODDR-UHFFFAOYSA-N
Synonym bismuth oxychloride, mearlite, pearl white, bismuthine, chlorooxo, wismutchloridoxid, bismut-chlorid-oxid, biju, bismuth chloride oxide, basic bismuth chloride, wismut iii-chlorid-oxid
PubChem CID 6328152
IUPAC Name oxobismuth;hydrochloride
SMILES O=[Bi].Cl

Specifications

Melting Point (decomposition)
Color Brown
Physical Form Powder
Quantity 10 g
Linear Formula BiOCl
Merck Index 14,1263
Formula Weight 260.43
Percent Purity 99.999%
Odor Odorless
Refractive Index 2.15
Density 7.72 g/mL
Assay (metals basis)
Chemical Name or Material Bismuth(III) chloride oxide
Show More Show Less
compliance-icons
Product Identifier
  • Bismuth(III) chloride oxide
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.