Learn More
Bismuth(III) chloride, ultra dry, 99.997+% (metals basis), Thermo Scientific Chemicals
100.00 € - 358.00 €
Chemical Identifiers
CAS | 7787-60-2 |
---|---|
Molecular Formula | BiCl3 |
Molecular Weight (g/mol) | 315.33 |
MDL Number | MFCD00003461 |
InChI Key | JHXKRIRFYBPWGE-UHFFFAOYSA-K |
Synonym | bismuth trichloride, bismuth chloride, trichlorobismuthine, trichlorobismuth, bismuth iii chloride, bismuthine, trichloro, trichlorobismute, bismuth lll chloride |
PubChem CID | 24591 |
IUPAC Name | trichlorobismuthane |
SMILES | Cl[Bi](Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11353847
|
Thermo Scientific Alfa Aesar
035680.06 |
5 g |
100.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11363847
|
Thermo Scientific Alfa Aesar
035680.14 |
25 g |
358.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bismuth chloride is used as a catalyst in organic synthesis. In particular, it catalyzes the Michael reaction and the Mukaiyama-aldol reaction. The addition of other metal iodides increases its catalytic activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Chemical Identifiers
7787-60-2 | |
315.33 | |
JHXKRIRFYBPWGE-UHFFFAOYSA-K | |
24591 | |
Cl[Bi](Cl)Cl |
BiCl3 | |
MFCD00003461 | |
bismuth trichloride, bismuth chloride, trichlorobismuthine, trichlorobismuth, bismuth iii chloride, bismuthine, trichloro, trichlorobismute, bismuth lll chloride | |
trichlorobismuthane |
Specifications
∼430°C (sublimation) | |
7787-60-2 | |
Powder | |
MFCD00003461 | |
14,1262 | |
It is soluble in methanol, diethyl ether, acetone. | |
Cl[Bi](Cl)Cl | |
315.33 | |
315.34 | |
Ampule | |
4.75 g/mL | |
Bismuth(III) chloride, Ultra dry |
230°C to 232°C | |
White | |
BiCl3 | |
UN3260 | |
bismuth trichloride, bismuth chloride, trichlorobismuthine, trichlorobismuth, bismuth iii chloride, bismuthine, trichloro, trichlorobismute, bismuth lll chloride | |
JHXKRIRFYBPWGE-UHFFFAOYSA-K | |
trichlorobismuthane | |
24591 | |
≥99.997% | |
Hygroscopic | |
(metals basis) |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 232-123-2
missing translation for 'rtecsNumber' : EB2690000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Ampouled under argon
Recommended Storage : Ambient temperatures; Store under Nitrogen
- Bismuth(III) chloride
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only