missing translation for 'onlineSavingsMsg'
Learn More
Bismuth(III) iodide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals Product Code.: 11373847

Bismuth(III) iodide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Product Code. 11373847
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11373847

Brand: Thermo Scientific Alfa Aesar 035681.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-64-6
Molecular Formula BiI3
Molecular Weight (g/mol) 589.69
MDL Number MFCD00010894
InChI Key KOECRLKKXSXCPB-UHFFFAOYSA-K
Synonym bismuth iodide, bismuth iii iodide, bismuth triiodide, bismuthine, triiodo, triiodobismuthine, triiodobismuth, bismuth iodide,, ksc179g5dShow MoreShow Less
PubChem CID 111042
IUPAC Name bismuth(3+) triiodide
SMILES [I-].[I-].[I-].[Bi+3]

Specifications

Boiling Point ∼500°C
Melting Point 408°C
Physical Form Granules
Quantity 5 g
UN Number UN3260
Merck Index 14,1269
Formula Weight 589.69
Percent Purity 99.998%
Odor Acidic
Packaging Ampule
Sensitivity Hygroscopic
Density 5.778 g/mL
Assay (metals basis)
Chemical Name or Material Bismuth(III) iodide, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bismuth(III) iodide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.