missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) nitrate pentahydrate, 98%, pure, Thermo Scientific Chemicals

Product Code. 15249916
Click to view available options
Quantity:
1 kg
5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-06-0
Molecular Formula BiH10N3O14
Molecular Weight (g/mol) 485.07
MDL Number MFCD00149157
InChI Key FBXVOTBTGXARNA-UHFFFAOYSA-N
Synonym bismuth nitrate pentahydrate, bismuth trinitrate pentahydrate, bismuth nitrate, hydrate 1:5, bismuth iii nitrate pentahydrate, unii-fo2icy167b, fo2icy167b, nitric acid, bismuth 3+ salt, pentahydrate, acmc-20alle, bismuthnitratepentahydrate
PubChem CID 160911
IUPAC Name bismuth;trinitrate;pentahydrate
SMILES O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Boiling Point 80.0°C
Melting Point 30.0°C
Quantity 5 kg
Assay Percent Range 97.5% min. (Complexometry)
Linear Formula Bi(NO3)3·5H2O
Merck Index 15,1271
Solubility Information Solubility in water: may decompose.
Formula Weight 485.07
Percent Purity 98%
Grade Pure
Chemical Name or Material Bismuth(III) nitrate pentahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Bismuth(III) nitrate pentahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'