Learn More
Bismuth(III) nitrate pentahydrate, ACS reagent, Thermo Scientific Chemicals
64.12 € - 208.04 €
Chemical Identifiers
CAS | 10035-06-0 |
---|---|
Molecular Formula | BiH10N3O14 |
Molecular Weight (g/mol) | 485.07 |
MDL Number | MFCD00149157 |
InChI Key | FBXVOTBTGXARNA-UHFFFAOYSA-N |
Synonym | bismuth nitrate pentahydrate, bismuth trinitrate pentahydrate, bismuth nitrate, hydrate 1:5, bismuth iii nitrate pentahydrate, unii-fo2icy167b, fo2icy167b, nitric acid, bismuth 3+ salt, pentahydrate, acmc-20alle, bismuthnitratepentahydrate |
PubChem CID | 160911 |
SMILES | O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
13418608
|
Thermo Scientific Acros
447561000 |
100 g | Plastic bottle |
64.12 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13428608
|
Thermo Scientific Acros
447565000 |
500 g | Plastic bottle |
208.04 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10035-06-0 | |
485.07 | |
FBXVOTBTGXARNA-UHFFFAOYSA-N | |
160911 |
BiH10N3O14 | |
MFCD00149157 | |
bismuth nitrate pentahydrate, bismuth trinitrate pentahydrate, bismuth nitrate, hydrate 1:5, bismuth iii nitrate pentahydrate, unii-fo2icy167b, fo2icy167b, nitric acid, bismuth 3+ salt, pentahydrate, acmc-20alle, bismuthnitratepentahydrate | |
O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
80.0°C | |
10035-06-0 | |
Bi(NO3)3·5H2O | |
15,1271 | |
Solubility in water: may decompose | |
O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
485.07 | |
485.07 | |
Bismuth(III) nitrate pentahydrate |
30.0°C | |
BiH10N3O14 | |
MFCD00149157 | |
bismuth nitrate pentahydrate, bismuth trinitrate pentahydrate, bismuth nitrate, hydrate 1:5, bismuth iii nitrate pentahydrate, unii-fo2icy167b, fo2icy167b, nitric acid, bismuth 3+ salt, pentahydrate, acmc-20alle, bismuthnitratepentahydrate | |
FBXVOTBTGXARNA-UHFFFAOYSA-N | |
bismuth(3+) pentahydrate trinitrate | |
160911 | |
ACS Reagent |
Safety and Handling
GHS Signal Word: Danger
- Bismuth(III) nitrate pentahydrate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H272-May intensify fire; oxidiser.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.