missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) oxide, 99.9%, (trace metal basis)

Product Code. 10451441 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10451441

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1304-76-3 | Bi2O3 | 465.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1304-76-3
Molecular Formula Bi2O3
Molecular Weight (g/mol) 465.96
MDL Number MFCD00003462
InChI Key TYIXMATWDRGMPF-UHFFFAOYSA-N
Synonym Bismuth trioxide
IUPAC Name dibismuth(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Bi+3].[Bi+3]

Specifications

Boiling Point 1890°C
Melting Point 825.0°C
Color Yellow
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.9%
Linear Formula Bi2O3
Fieser 01,59
Merck Index 15,1273
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acids
Formula Weight 465.95
Percent Purity 99.9%
Grade Extra Pure
Loss on Drying 0.2% max.
Packaging Plastic bottle
Specific Gravity 8.9
Density 8.9000g/mL
Chemical Name or Material Bismuth(III) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.