missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth(III) oxide, NanoArc™, 99.5+%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
500 g
Description
Bismuth(III) oxide is used in solid-oxide fuel cells. It is also used in optical glasses, to replace lead oxide in whitewares, luxes, varistor formulations and ceramic capacitor formulations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- X-ray opaque particle
- NanoArc™ bismuth oxide can be compounded into polymers or coating systems to increase opacity to x-rays,while the small particle size ensures minimal disruption of physical properties
Spécification
Spécification
Boiling Point | ∼1890°C |
Melting Point | 817°C |
Color | Yellow |
Physical Form | Powder |
Quantity | 25 g |
Merck Index | 14,1273 |
Solubility Information | Soluble in acids. Insoluble in water. |
Formula Weight | 465.96 |
Percent Purity | ≥99.5% |
Odor | Odorless |
Afficher plus |
RUO – Research Use Only
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Bismuth(III) oxide, NanoArc™, 99.5+%, Thermo Scientific Chemicals >
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu