Learn More
Bismuth(III) oxyfluoride, 97%, pure, Thermo Scientific™
67.00 € - 425.00 €
Chemical Identifiers
CAS | 13520-72-4 |
---|---|
Molecular Formula | BiFO |
Molecular Weight (g/mol) | 243.98 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15249856
|
Thermo Scientific Acros
453820250 |
25 g | N/A | ||||||
15259856
|
Thermo Scientific Acros
453821000 |
100 g |
186.93 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15249866
|
Thermo Scientific Acros
453825000 |
500 g | N/A | ||||||
Description
Chemical Identifiers
13520-72-4 | |
BiFO | |
243.98 |
Specifications
500.0°C | |
96% min. (ex Bi) | |
FBi=O | |
243.98 | |
97% | |
Bismuth(III) oxyfluoride |
13520-72-4 | |
BiFO | |
Solubility in water: insoluble. | |
243.98 | |
Pure |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep only in
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 236-861-6
- Bismuth(III) oxyfluoride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H290-May be corrosive to metals.
Precautionary Statement
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P390-Absorb spillage to prevent material damage.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.