missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) phosphate, 99%, Thermo Scientific Chemicals

92.96 € - 322.61 €

Chemical Identifiers

CAS 10049-01-1
Molecular Formula BiH3O4P
Molecular Weight (g/mol) 306.97
MDL Number MFCD00049721
InChI Key SFOQXWSZZPWNCL-UHFFFAOYSA-N
Synonym bismuth phosphate, bismuth iii phosphate, bismugel, phosphoric acid, bismuth 3+ salt 1:1, bismuth phosphate mi, bismuth phosphate who-dd, acmc-20allc, bismuth orthophosphate, bi.po4, bismuth phosphate,
PubChem CID 9839429
SMILES [Bi+3].OP(O)(O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11338158
View Documents
Thermo Scientific Alfa Aesar
039380.18
50 g
92.96 €
50g
Please to purchase this item. Need a web account? Register with us today!
11328158
View Documents
Thermo Scientific Alfa Aesar
039380.30
250 g
322.61 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Bismuth(III) phosphate is used in treatment of renal cysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10049-01-1
306.97
SFOQXWSZZPWNCL-UHFFFAOYSA-N
9839429
BiH3O4P
MFCD00049721
bismuth phosphate, bismuth iii phosphate, bismugel, phosphoric acid, bismuth 3+ salt 1:1, bismuth phosphate mi, bismuth phosphate who-dd, acmc-20allc, bismuth orthophosphate, bi.po4, bismuth phosphate,
[Bi+3].OP(O)(O)=O

Specifications

(decomposition)
Powder
MFCD00049721
bismuth phosphate, bismuth iii phosphate, bismugel, phosphoric acid, bismuth 3+ salt 1:1, bismuth phosphate mi, bismuth phosphate who-dd, acmc-20allc, bismuth orthophosphate, bi.po4, bismuth phosphate,
SFOQXWSZZPWNCL-UHFFFAOYSA-N
bismuth;phosphate
9839429
99%
6.32 g/mL
10049-01-1
BiH3O4P
14,1275
Insoluble in water and alkali; Soluble in hydrochloric acid
[Bi+3].OP(O)(O)=O
306.97
303.95
Odorless
Bismuth(III) phosphate
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 233-161-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.